2,3-Difluoro-4-(methoxymethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

57366-1G 57366-25G 57366-500MG 57366-5G
76892-044EA 266.29 USD
76892-044 76892-046 76892-048 76892-050
2,3-Difluoro-4-(methoxymethoxy)benzoic acid ≥97%
2,3-Difluoro-4-(methoxymethoxy)benzoic acid
Formula: C₉H₈F₂O₄
MW: 218.154 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34530591
CAS Number: 2734777-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR