1-Bromo-4-chloro-2,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2,3-difluorobromobenzene

57297-1G 57297-25G 57297-5G
76892-030EA 114.58 USD
76892-030 76892-032 76892-034
1-Bromo-4-chloro-2,3-difluorobenzene ≥97%
1-Bromo-4-chloro-2,3-difluorobenzene
Formula: C₆H₂BrClF₂
MW: 227.434 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878201
CAS Number: 1000574-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR