1-Isobutyl-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

57258-1G 57258-25G 57258-500MG 57258-5G
76892-014EA 411.63 USD
76892-014 76892-016 76892-018 76892-020
1-Isobutyl-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
1-Isobutyl-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Formula: C₁₀H₁₆N₂O₂
MW: 196.246 g/mol
Storage Temperature: Ambient
CAS Number: 1017486-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR