1-Isopropyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

57251-1G 57251-25G 57251-5G
76892-002EA 62.6 USD
76892-002 76892-004 76892-006
1-Isopropyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde ≥97%
1-Isopropyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde
Formula: C₉H₁₄N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD06260699
CAS Number: 890626-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR