2-(2-Chloropyridin-4-yl)acetic acid ≥97%

Supplier: AOBChem USA

Warning

57126-25G 57126-5G
76891-964EA 605.78 USD
76891-964 76891-966
2-(2-Chloropyridin-4-yl)acetic acid ≥97%
2-(2-Chloropyridin-4-yl)acetic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367936
CAS Number: 887580-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR