1-(2,4-Dichloro-3-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

56837-25G 56837-5G
76891-952EA 563.34 USD
76891-952 76891-954
1-(2,4-Dichloro-3-methylphenyl)ethanone ≥97%
1-(2,4-Dichloro-3-methylphenyl)ethanone
Formula: C₉H₈Cl₂O
MW: 203.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28128939
CAS Number: 157652-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR