2-Bromo-6-iodoanisole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-3-iodo-2-methoxybenzene, 2-Bromo-6-iodophenyl methyl ether

56778-1G 56778-25G 56778-5G
76891-938EA 114.58 USD
76891-938 76891-940 76891-942
2-Bromo-6-iodoanisole ≥95%
2-Bromo-6-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆BrIO
Storage Temperature: Ambient
MDL Number: MFCD18391617
CAS Number: 860584-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR