(1-Phenyl-1H-pyrazol-5-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

56564-1G 56564-25G 56564-500MG 56564-5G
76891-922EA 434.97 USD
76891-922 76891-924 76891-926 76891-928
(1-Phenyl-1H-pyrazol-5-yl)methanol ≥97%
(1-Phenyl-1H-pyrazol-5-yl)methanol
Formula: C₁₀H₁₀N₂O
MW: 174.199 g/mol
Storage Temperature: Ambient
CAS Number: 1017783-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR