2-Isopropyl-2H-pyrazole-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Isopropyl-1H-pyrazole-5-carbaldehyde

56552-1G 56552-25G 56552-5G
76891-902EA 114.58 USD
76891-902 76891-904 76891-906
2-Isopropyl-2H-pyrazole-3-carbaldehyde ≥97%
2-Isopropyl-2H-pyrazole-3-carbaldehyde
Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03130042
CAS Number: 100305-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR