2,4-Dichloro-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

56465-1G 56465-500MG 56465-5G
76891-888EA 970.73 USD
76891-888 76891-890 76891-892
2,4-Dichloro-5-formylbenzoic acid ≥97%
2,4-Dichloro-5-formylbenzoic acid
Formula: C₈H₄Cl₂O₃
MW: 219.022 g/mol
Storage Temperature: Ambient
CAS Number: 1936558-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR