(3,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

56115-1G 56115-25G 56115-500MG 56115-5G
76891-860EA 228.1 USD
76891-860 76891-862 76891-864 76891-866
(3,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol ≥97% research grade
(3,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol
Formula: C₁₃H₁₇BF₂O₃
Storage Temperature: Ambient
CAS Number: 1857323-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR