1-Bromo-4-iodo-2-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

55977-1G 55977-25G 55977-500MG 55977-5G
76891-844EA 263.11 USD
76891-844 76891-846 76891-848 76891-850
1-Bromo-4-iodo-2-methoxy-3-methylbenzene ≥97%
1-Bromo-4-iodo-2-methoxy-3-methylbenzene
Formula: C₈H₈BrIO
Storage Temperature: Refrigerator
CAS Number: 2385950-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR