2-(1-Hydroxycyclobutyl)acetic acid ≥95%

Supplier: AOBChem USA

Warning

55579-1G 55579-25G 55579-500MG 55579-5G
76891-818EA 441.34 USD
76891-818 76891-820 76891-822 76891-824
2-(1-Hydroxycyclobutyl)acetic acid ≥95%
2-(1-Hydroxycyclobutyl)acetic acid
Formula: C₆H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD16749170
CAS Number: 933695-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR