1-Fluoro-4-iodo-3-methoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

55495-1G 55495-25G 55495-500MG 55495-5G
76891-802EA 292.81 USD
76891-802 76891-804 76891-806 76891-808
1-Fluoro-4-iodo-3-methoxy-2-methylbenzene ≥97%
1-Fluoro-4-iodo-3-methoxy-2-methylbenzene
Formula: C₈H₈FIO
Storage Temperature: Ambient
MDL Number: MFCD34564302
CAS Number: 2387149-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR