1-Bromo-2,4-dimethoxy-5-fluorobenzene 97%

Supplier: AOBChem USA

Warning

55466-1G 55466-25G 55466-5G 55466-500MG
76891-788EA 117.76 USD
76891-788 76891-790 76891-792 77651-636
1-Bromo-2,4-dimethoxy-5-fluorobenzene 97%
1-Bromo-2,4-dimethoxy-5-fluorobenzene
Formula: C₈H₈BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12547863
CAS Number: 1208077-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR