5-Hydroxy-2-(methylthio)pyrimidine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Methylthio)pyrimidin-5-ol

54957-1G 54957-25G 54957-500MG 54957-5G
76891-728EA 146.41 USD
76891-728 76891-730 76891-732 76891-734
5-Hydroxy-2-(methylthio)pyrimidine ≥97%
5-Hydroxy-2-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₆N₂OS
MW: 142.179 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00223737
CAS Number: 4874-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR