1-Chloro-2-fluoro-4-iodo-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

54904-1G 54904-25G 54904-500MG 54904-5G
76891-698EA 233.4 USD
76891-698 76891-700 76891-702 76891-704
1-Chloro-2-fluoro-4-iodo-5-methylbenzene ≥95%
1-Chloro-2-fluoro-4-iodo-5-methylbenzene
Formula: C₇H₅ClFI
MW: 270.47 g/mol
Storage Temperature: Ambient
CAS Number: 1805502-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR