1-Bromo-3-fluoro-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

54845-1G 54845-25G 54845-500MG 54845-5G
76891-684EA 197.33 USD
76891-684 76891-686 76891-688 76891-690
1-Bromo-3-fluoro-2,5-dimethylbenzene ≥97%
1-Bromo-3-fluoro-2,5-dimethylbenzene
Formula: C₈H₈BrF
MW: 203.052 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28609392
CAS Number: 1780924-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR