2,4,5-Trichlorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

54814-1G 54814-25G 54814-500MG 54814-5G
76891-658EA 363.89 USD
76891-658 76891-660 76891-662 76891-664
2,4,5-Trichlorobenzaldehyde ≥95%
2,4,5-Trichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₃O
Storage Temperature: Ambient
MDL Number: MFCD09999820
CAS Number: 35696-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR