2'-Fluoro-5'-methylacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2-Fluoro-5-methylphenyl)ethanone

54757-25G 54757-5G
76891-652EA 257.8 USD
76891-652 76891-654
2'-Fluoro-5'-methylacetophenone ≥97%
2'-Fluoro-5'-methylacetophenone

Precaución: for Research Use Only

Formula: C₉H₉FO
MW: 152.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236680
CAS Number: 446-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR