2-(6-Isobutoxy-naphthalen-2-yl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

54461-1G 54461-25G 54461-5G 54461-500MG
76891-602EA 84.88 USD
76891-602 76891-604 76891-606 77647-584
2-(6-Isobutoxy-naphthalen-2-yl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane ≥97%
2-(6-Isobutoxy-naphthalen-2-yl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane

Precaución: for Research Use Only

Formula: C₂₀H₂₇BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD26142761
CAS Number: 1363386-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR