1-(Methoxymethoxy)-2,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

54405-25G 54405-5G
76891-590EA 204.76 USD
76891-590 76891-592
1-(Methoxymethoxy)-2,3-dimethylbenzene ≥97%
1-(Methoxymethoxy)-2,3-dimethylbenzene
Formula: C₁₀H₁₄O₂
MW: 166.217 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31376087
CAS Number: 1694512-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR