1-Chloro-3-(methoxymethoxy)-2,4-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

54281-1G 54281-25G 54281-500MG 54281-5G
76891-574EA 171.87 USD
76891-574 76891-576 76891-578 76891-580
1-Chloro-3-(methoxymethoxy)-2,4-dimethylbenzene ≥97%
1-Chloro-3-(methoxymethoxy)-2,4-dimethylbenzene
Formula: C₁₀H₁₃ClO₂
Storage Temperature: Refrigerator
CAS Number: 2921833-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR