4-Bromo-2-chloro-5-methylanisole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-5-chloro-4-methoxy-2-methylbenzene

54147-1G 54147-25G 54147-5G
76891-562EA 79.57 USD
76891-562 76891-564 76891-566
4-Bromo-2-chloro-5-methylanisole ≥95%
4-Bromo-2-chloro-5-methylanisole

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Refrigerator
MDL Number: MFCD11504949
CAS Number: 179897-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR