1-Bromo-5-chloro-2,4-difluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

53856-1G 53856-25G 53856-5G 53856-500MG
76891-520EA 88.06 USD
76891-520 76891-522 76891-524 77647-574
1-Bromo-5-chloro-2,4-difluoro-3-iodobenzene ≥97%
1-Bromo-5-chloro-2,4-difluoro-3-iodobenzene
Formula: C₆HBrClF₂I
MW: 353.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530232
CAS Number: 2292219-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR