1-Fluoro-2,4,5-trimethoxybenzene 95%

Supplier: AOBChem USA

Warning

53764-1G 53764-500MG 53764-5G 53764-25G
76891-496EA 761.73 USD
76891-496 76891-498 76891-500 77656-456
1-Fluoro-2,4,5-trimethoxybenzene 95%
1-Fluoro-2,4,5-trimethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO₃
MW: 186.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34757130
CAS Number: 2385965-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR