1-Bromo-5-fluoro-2-methoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

53541-1G 53541-500MG 53541-5G
76891-474EA 672.62 USD
76891-474 76891-476 76891-478
1-Bromo-5-fluoro-2-methoxy-4-methylbenzene ≥97%
1-Bromo-5-fluoro-2-methoxy-4-methylbenzene
Formula: C₈H₈BrFO
Storage Temperature: Ambient
CAS Number: 1783385-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR