1-(3-Chloro-2-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

53473-1G 53473-25G 53473-500MG 53473-5G
76891-454EA 173.99 USD
76891-454 76891-456 76891-458 76891-460
1-(3-Chloro-2-methoxyphenyl)ethanone ≥97%
1-(3-Chloro-2-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08753704
CAS Number: 99585-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR