(2-Bromo-3-fluorophenoxy)(tert-butyl)dimethylsilane ≥95%

Supplier: AOBChem USA

Warning

53229-1G 53229-25G 53229-500MG 53229-5G
76891-424EA 233.4 USD
76891-424 76891-426 76891-428 76891-430
(2-Bromo-3-fluorophenoxy)(tert-butyl)dimethylsilane ≥95%
(2-Bromo-3-fluorophenoxy)(tert-butyl)dimethylsilane
Formula: C₁₂H₁₈BrFOSi
MW: 305.259 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29043963
CAS Number: 1881332-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR