2-(Benzyloxy)-4-chlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53149-1G 53149-25G 53149-500MG 53149-5G
76891-384EA 292.81 USD
76891-384 76891-386 76891-388 76891-390
2-(Benzyloxy)-4-chlorobenzaldehyde ≥97%
2-(Benzyloxy)-4-chlorobenzaldehyde
Formula: C₁₄H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD20127036
CAS Number: 1334615-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR