3-Bromo-5-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-3-iodo-5-methoxybenzene

53111-1G 53111-25G 53111-500MG 53111-5G
76891-370EA 141.1 USD
76891-370 76891-372 76891-374 76891-376
3-Bromo-5-iodoanisole ≥97%
3-Bromo-5-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆BrIO
Storage Temperature: Refrigerator
MDL Number: MFCD12547766
CAS Number: 915412-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR