1-Ethoxy-2-fluoro-4-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

53048-1G 53048-25G 53048-500MG 53048-5G
76891-330EA 263.11 USD
76891-330 76891-332 76891-334 76891-336
1-Ethoxy-2-fluoro-4-iodobenzene ≥95%
1-Ethoxy-2-fluoro-4-iodobenzene
Formula: C₈H₈FIO
MW: 266.051 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12547860
CAS Number: 1208075-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR