6-Chloro-2-fluoro-3-methylbenzyl alcohol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (6-Chloro-2-fluoro-3-methylphenyl)methanol

52858-1G 52858-25G 52858-5G
76891-318EA 66.84 USD
76891-318 76891-320 76891-322
6-Chloro-2-fluoro-3-methylbenzyl alcohol ≥97% research grade
6-Chloro-2-fluoro-3-methylbenzyl alcohol
Formula: C₈H₈ClFO
MW: 174.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01631372
CAS Number: 261762-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR