1-Bromo-4-chloro-2-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52823-1G 52823-25G 52823-5G
76891-302EA 289.63 USD
76891-302 76891-304 76891-308
1-Bromo-4-chloro-2-methoxy-5-methylbenzene ≥97%
1-Bromo-4-chloro-2-methoxy-5-methylbenzene
Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Refrigerator
CAS Number: 83803-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR