3,4-Dimethoxythioanisole ≥95%

Supplier: AOBChem USA

Warning

52734-25G 52734-5G
76891-296EA 484.84 USD
76891-296 76891-298
3,4-Dimethoxythioanisole ≥95%
3,4-Dimethoxythioanisole

Precaución: for Research Use Only

Formula: C₉H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD03789172
CAS Number: 2570-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR