(5-Bromo-2-Iodophenyl)(Methyl)Sulfane ≥97%

Supplier: AOBChem USA

Warning

52583-1G 52583-25G 52583-5G
76891-284EA 64.72 USD
76891-284 76891-286 76891-288
(5-Bromo-2-Iodophenyl)(Methyl)Sulfane ≥97%
(5-Bromo-2-Iodophenyl)(Methyl)Sulfane
Formula: C₇H₆BrIS
MW: 328.996 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22489402
CAS Number: 1824602-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR