1-Bromo-2-fluoro-3-iodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

52566-1G 52566-25G 52566-5G 52566-500MG
76891-270EA 88.06 USD
76891-270 76891-272 76891-274 77647-554
1-Bromo-2-fluoro-3-iodo-4-methoxybenzene ≥97%
1-Bromo-2-fluoro-3-iodo-4-methoxybenzene
Formula: C₇H₅BrFIO
MW: 330.921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723577
CAS Number: 1628450-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR