1,2-Dibromo-4-ethylbenzene ≥95%

Supplier: AOBChem USA

Warning

52535-1G 52535-25G 52535-5G
76891-256EA 84.88 USD
76891-256 76891-258 76891-260
1,2-Dibromo-4-ethylbenzene ≥95%
1,2-Dibromo-4-ethylbenzene
Formula: C₈H₈Br₂
Storage Temperature: Refrigerator
MDL Number: MFCD00673000
CAS Number: 134940-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR