1-Bromo-4-chloro-3-iodo-2-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

52501-1G 52501-25G 52501-500MG 52501-5G
76891-240EA 263.11 USD
76891-240 76891-242 76891-244 76891-246
1-Bromo-4-chloro-3-iodo-2-methoxybenzene ≥95%
1-Bromo-4-chloro-3-iodo-2-methoxybenzene
Formula: C₇H₅BrClIO
MW: 347.375 g/mol
Storage Temperature: Ambient
CAS Number: 1823903-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR