3,5-Bis(trifluoromethyl)toluene 95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Methyl-3,5-bis(trifluoromethyl)benzene

52260-1G 52260-25G 52260-5G 52260-500MG
76891-214EA 105.03 USD
76891-214 76891-216 76891-218 77651-534
3,5-Bis(trifluoromethyl)toluene 95%
3,5-Bis(trifluoromethyl)toluene

Precaución: for Research Use Only

Formula: C₉H₆F₆
Storage Temperature: Ambient
MDL Number: MFCD04972861
CAS Number: 75462-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR