(4-Bromo-2-chlorophenyl)(morpholino)methanone 97%

Supplier: AOBChem USA

Warning

52244-1G 52244-25G 52244-5G 52244-500MG
76891-204EA 96.55 USD
76891-204 76891-206 76891-208 77651-532
(4-Bromo-2-chlorophenyl)(morpholino)methanone 97%
(4-Bromo-2-chlorophenyl)(morpholino)methanone
Formula: C₁₁H₁₁BrClNO₂
MW: 304.568 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756426
CAS Number: 877383-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR