1H-Pyrazolo[3,4-c]pyridine ≥97%

Supplier: AOBChem USA

Warning

52158-25G 52158-5G
76891-194EA 254.62 USD
76891-194 76891-196
1H-Pyrazolo[3,4-c]pyridine ≥97%
1H-Pyrazolo[3,4-c]pyridine
Formula: C₆H₅N₃
MW: 119.1240 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00661308
CAS Number: 271-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR