1-Bromo-5-chloro-2-fluoro-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

51969-1G 51969-25G 51969-500MG 51969-5G
76891-170EA 183.54 USD
76891-170 76891-172 76891-174 76891-176
1-Bromo-5-chloro-2-fluoro-3-methylbenzene ≥95%
1-Bromo-5-chloro-2-fluoro-3-methylbenzene
Formula: C₇H₅BrClF
MW: 223.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846002
CAS Number: 146948-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR