5'-Chloro-2'-fluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(5-Chloro-2-fluorophenyl)ethanone

51963-25G 51963-5G
76891-158EA 598.35 USD
76891-158 76891-160
5'-Chloro-2'-fluoroacetophenone ≥97%
5'-Chloro-2'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MW: 172.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094519
CAS Number: 541508-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR