2,3-Dichloro-6-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

51914-25G 51914-5G
76891-150EA 310.85 USD
76891-150 76891-152
2,3-Dichloro-6-fluorobenzamide ≥97%
2,3-Dichloro-6-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₄Cl₂FNO
Storage Temperature: Ambient
MDL Number: MFCD04115877
CAS Number: 886497-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR