(4-Phenylthiophen-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

51691-1G 51691-25G 51691-500MG 51691-5G
76891-138EA 203.7 USD
76891-138 76891-140 76891-142 76891-144
(4-Phenylthiophen-2-yl)methanol ≥97%
(4-Phenylthiophen-2-yl)methanol
Formula: C₁₁H₁₀OS
MW: 190.262 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06202842
CAS Number: 79757-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR