2-(Cyclopropylmethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

51500-1G 51500-25G 51500-500MG 51500-5G
76891-110EA 185.66 USD
76891-110 76891-112 76891-114 76891-116
2-(Cyclopropylmethoxy)benzaldehyde ≥97%
2-(Cyclopropylmethoxy)benzaldehyde
Formula: C₁₁H₁₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11156430
CAS Number: 54280-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR