1-Chloro-4-iodo-2-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

51484-1G 51484-25G 51484-500MG 51484-5G
76891-096EA 155.96 USD
76891-096 76891-098 76891-100 76891-102
1-Chloro-4-iodo-2-(trifluoromethoxy)benzene ≥97%
1-Chloro-4-iodo-2-(trifluoromethoxy)benzene
Formula: C₇H₃ClF₃IO
MW: 322.451 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25956425
CAS Number: 1198422-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR