(4-Isopropoxy-3-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

51445-1G 51445-25G 51445-500MG 51445-5G
76891-064EA 233.4 USD
76891-064 76891-066 76891-068 76891-070
(4-Isopropoxy-3-methoxyphenyl)methanol ≥95%
(4-Isopropoxy-3-methoxyphenyl)methanol
Formula: C₁₁H₁₆O₃
MW: 196.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09754745
CAS Number: 2539-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR