2-(6-Isopropoxynaphthalen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade

Supplier: AOBChem USA

Warning

51419-1G 51419-25G 51419-500MG 51419-5G
76891-048EA 155.96 USD
76891-048 76891-050 76891-052 76891-054
2-(6-Isopropoxynaphthalen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade
2-(6-Isopropoxynaphthalen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₉H₂₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD30288517
CAS Number: 2246680-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR